missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Trastuzumab Antibody (2261E) [Allophycocyanin/Cy7], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals™ FAB95471APCCY7
This item is not returnable.
View return policy
Description
Trastuzumab Monoclonal antibody specifically detects Trastuzumab in Broad Species Reactivity samples. It is validated for Flow Cytometry
Specifications
| Trastuzumab | |
| Monoclonal | |
| APC-Cyanine7 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Broad Species Reactivity | |
| Purified |
| Flow Cytometry | |
| 2261E | |
| PBS | |
| Full-length Trastuzumab | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction