Learn More
Trastuzumab Mouse, Unconjugated, Monoclonal Antibody, AcroBiosystems
Product Code. 16498119
Change view
Click to view available options
Quantity:
1 mg
100 μg
Unit Size:
100µg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16498119 100 μg 100µg
16408129 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16498119 Supplier ACROBiosystems Supplier No. TRBY5b100ug
Mouse Monoclonal Antibody
Trastuzumab Monoclonal antibody specifically detects Trastuzumab.
Specifications
Antigen Trastuzumab
Classification Monoclonal
Conjugate Unconjugated
Host Species Human (HEK293)
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage -20°C