Learn More
Edmund Bühler GmbH™ Tray WS/TL
Click to view available options
Dimensions (L x W):
340 x 340 mm
340 x 500 mm
Unit Size:
Each
Description
Tray WS/TL as additional tray for multi-storey rack WS / TL incl. rubber mat
Specifications
Specifications
| Product Type | Tray |
| Dimensions (L x W) | 340 x 340 mm |
| For Use With (Equipment) | Multi-story rack system WS / TL |
| Includes | Rubber mat |