missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CA19-9/Sialyl Lewis A Antibody (1031516), PE/Cy5.5, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB10625PECY55
This item is not returnable.
View return policy
Description
CA19-9/Sialyl Lewis A Monoclonal antibody specifically detects CA19-9/Sialyl Lewis A in Human samples. It is validated for Flow Cytometry
Specifications
| CA19-9/Sialyl Lewis A | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| PBS | |
| Human CA19-9 from Capan-2 human pancreatic adenocarcinoma cell line | |
| 0.1 mL | |
| Tumor Biomarkers | |
| Human | |
| Purified |
| Flow Cytometry | |
| 1031516 | |
| Flow Cytometry | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Store at 4C in the dark. Do not freeze. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction