missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRBC1 Antibody (JOVI.1), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50355MFV450
This item is not returnable.
View return policy
Description
TRBC1 Monoclonal antibody specifically detects TRBC1 in Human samples. It is validated for Flow Cytometry
Specifications
| TRBC1 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| JOVI.1 | |
| Flow Cytometry | |
| BV05S1J2.2, TCRBC1, TRBC1 T cell receptor beta constant 1 | |
| Human HA1.7 TCR beta chain expressed on transgenic mouse cells. | |
| 0.1 mL | |
| Adaptive Immunity, Cell Biology, Diabetes Research, Immunology, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
| 28639 | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction