missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Treacher Collins syndrome protein Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP1-86909
This item is not returnable.
View return policy
Description
Treacher Collins syndrome protein Polyclonal specifically detects Treacher Collins syndrome protein in Human, Zebrafish samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Treacher Collins syndrome protein | |
| Polyclonal | |
| Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| MFD1, nucleolar trafficking phosphoprotein, TCS1, Treacher Collins syndrome protein, Treacher Collins-Franceschetti syndrome 1, treacle, treacle protein | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| TCOF1 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:ESSRISDGKKQEGPATQVSKKNPASLPLTQAALKVLAQKASEAQPPVARTQPSSGVDSAVGTLPATSPQSTSVQAKGTNKLRKPK | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 6949 | |
| Human, Zebrafish | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction