missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: LYQDDKQFVD MPLSIAPEQV LQTFTELSRD HNHSIPREQL QAFVHEHFQA KGQELQPWTP ADWKDSPQFL QKISDAKLRA WAGQLHQLWK KLGKK Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 73%.
This gene encodes an enzyme that hydrolyses trehalose, a disaccharide formed from two glucose molecules found mainly in fungi, plants, and insects. A partial duplication of this gene is located adjacent to this locus on chromosome 11.
Specifications
Specifications
| Antigen | TREH |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | TREH |
| Gene Accession No. | O43280 |
| Gene Alias | 2210412M19Rik; Alpha,alpha-trehalase; alpha,alpha-trehalose glucohydrolase; LOW QUALITY PROTEIN: trehalase; trehalase; si:ch211-147p17.2; TRE; TREA; TREH; Trehalase; trehalase (brush-border membrane glycoprotein) |
| Gene Symbols | TREH |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
