missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TREM1 Antibody (CL7111) [Janelia Fluor« 525], Novus Biologicals Biologicals™
Shop All Bio Techne ProductsDescription
TREM1 Monoclonal antibody specifically detects TREM1 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | TREM1 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | CL7111 |
| Conjugate | Janelia Fluor 525 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | CD354 antigen, TREM-1Triggering receptor expressed on monocytes 1, triggering receptor expressed on myeloid cells 1, triggering-receptor TREM1 |
| Host Species | Mouse |
| Immunogen | This antibody was generated using a synthetic peptide of Q9NP99, with the exact immunogen sequence remaining proprietary. |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?