Learn More
TREM1 Antibody (CL7111) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-76531JF669
Product Code. 30307584
  • € 737.00 / 0.10mL
Description
TREM1 Monoclonal antibody specifically detects TREM1 in Human samples. It is validated for Western Blot,KnockDown
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TREM1
Monoclonal
Janelia Fluor 669
CD354 antigen, TREM-1Triggering receptor expressed on monocytes 1, triggering receptor expressed on myeloid cells 1, triggering-receptor TREM1
This antibody was developed using a synthetic peptide derived from Q9NP99, with the exact immunogen sequence remaining proprietary.
0.1 mL
Signal Transduction
54210
Store at 4⪚C in the dark.
IgG1
Western Blot, KnockDown
CL7111
50mM Sodium Borate
Mouse
Protein A purified