missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TREM1 Antibody (MM0583-9T3), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-11977AF594
This item is not returnable.
View return policy
Description
TREM1 Monoclonal specifically detects TREM1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| TREM1 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| TREM1 | |
| Human recombinant TREM1 (extracellular domain) | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| MM0583-9T3 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| CD354 antigen, TREM-1Triggering receptor expressed on monocytes 1, triggering receptor expressed on myeloid cells 1, triggering-receptor TREM1 | |
| Mouse | |
| 0.1 mL | |
| 54210 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction