missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TREM2b Antibody (RM0139-5J46), Alexa Fluor™ 700, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-22515AF700
This item is not returnable.
View return policy
Description
TREM2b Monoclonal specifically detects TREM2b in Mouse samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| TREM2b | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| TREM2 | |
| Mouse recombinant TREM2b (extracellular domain) | |
| 0.1 mL | |
| 54209 | |
| Mouse | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry | |
| RM0139-5J46 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry | |
| Trem2a, Trem2b, Trem2c, TREM-2triggering receptor expressed on myeloid cells 2a, Triggering receptor expressed on monocytes 2, triggering receptor expressed on myeloid cells 2 | |
| Rat | |
| Protein A or G purified | |
| Primary | |
| This antibody detects mouse of TREM-2b, but not TREM-1 in Western blotting. | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction