missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TREML2 Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA547833
This item is not returnable.
View return policy
Description
In direct ELISAs, less than 1% cross-reactivity with recombinant mouse (rm) TREML1, rmTREML4, rmTREM1, rmTREM2b, rmTREM3, rmTREM4, and recombinant human TREM2 is observed. Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.
TREML2 is located in a gene cluster on chromosome 6 with the single Ig variable (IgV) domain activating receptors TREM1 and TREM2, but it has distinct structural and functional properties.
Specifications
| TREML2 | |
| Polyclonal | |
| Unconjugated | |
| TREML2 | |
| AW049306; C6orf76; dJ238O23.1; FLJ13693; Gm750; MGC149715; MGC149716; Tlt2; TLT-2; TREM; Treml2; Trem-like transcript 2 protein; Triggering receptor expressed on myeloid cells; triggering receptor expressed on myeloid cells like 2; triggering receptor expressed on myeloid cells-like 2; triggering receptor expressed on myeloid cells-like 2a dominant; triggering receptor expressed on myeloid cells-like protein 2; UNQ6268/PRO20473 | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 328833 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q2LA85 | |
| TREML2 | |
| CHO-derived recombinant mouse TREML2/TLT-2 His25-Ala270. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction