missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TREML2/TLT-2 Antibody (001), FITC , Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89948F
This item is not returnable.
View return policy
Description
TREML2/TLT-2 Monoclonal antibody specifically detects TREML2/TLT-2 in Human samples. It is validated for ELISA
Specifications
| TREML2/TLT-2 | |
| Monoclonal | |
| FITC | |
| C6orf76, dJ238O23.1, FLJ13693, MGC149715, MGC149716, TLT2, TLT-2, trem-like transcript 2 protein, triggering receptor expressed on myeloid cells-like 2, triggering receptor expressed on myeloid cells-like protein 2, UNQ6268/PRO20473 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human TREML2/TLT-2 (NP_079083.2; Met1-Ser268). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 001 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 79865 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction