missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TREML2/TLT-2 Antibody (656906), mFluor Violet 610 SE, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB6707MFV610
This item is not returnable.
View return policy
Description
TREML2/TLT-2 Monoclonal antibody specifically detects TREML2/TLT-2 in Mouse samples. It is validated for Flow Cytometry, CyTOF
Specifications
| TREML2/TLT-2 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 79865 | |
| Store at 4C in the dark. | |
| IgG2a |
| Flow Cytometry, CyTOF | |
| 656906 | |
| Flow Cytometry, CyTOF-ready | |
| C6orf76, dJ238O23.1, FLJ13693, MGC149715, MGC149716, TLT2, TLT-2, trem-like transcript 2 protein, triggering receptor expressed on myeloid cells-like 2, triggering receptor expressed on myeloid cells-like protein 2, UNQ6268/PRO20473 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse TREML2/TLT-2, His25-Ala270, Accession # Q2LA85 | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction