missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Treponema pallidum p47 Monoclonal Antibody (2125)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA533298

Product Code. 17175193

  • € 528.00 / 1mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Does not cross react with Borrelia burgdorferi. Does not stain the entire bacterial cell.

Treponema pallidum is a gram-negative spirochaete bacterium and is considered to be metabolically crippled. There are at least four known subspecies: T. pallidum pallidum, T. pallidum pertenue, T. pallidum carateum and T. pallidum endemicum. The helical structure of T. pallidum pallidum allows it to move in a corkscrew motion through viscous mediums such as mucus. Treponema pallidum subsp. pallidum has one of the smallest bacterial genomes at 1.14 million base pairs (Mb) and has limited metabolic capabilities, reflecting its adaptation through genome reduction to the rich environment of mammalian tissue.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Treponema pallidum p47
Monoclonal
100 μg/mL
PBS with 0.1% sodium azide; pH 7.2
Mouse
1 mL
Primary
4°C
Liquid
ELISA, Western Blot, Immunocytochemistry
2125
Unconjugated
Syphillis; t pallidum; T. pallidum
Protein A
RUO
Bacteria
Antibody
IgG2b
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.