missing translation for 'onlineSavingsMsg'
Learn More

Tretazicar (>94%), TRC

Product Code. 30457272
Change view
Click to view available options
Quantity:
10 mg
50 mg
500 mg
Unit Size:
10mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30457272 50 mg -
30449940 10 mg 10mg
30451166 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30457272 Supplier Toronto Research Chemicals Supplier No. TRCT71980050MG
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21919-05-1
Molecular Formula C9H8N4O5
Molecular Weight (g/mol) 252.18
Synonym 5-(1-Aziridinyl)-2,4-dinitrobenzamide, 2,4-Dinitro-5-ethyleneiminobenzamide, 2,4-Dinitroethyleneiminobenzamide, 5-(1-Aziridinyl)-2,4-dinitrobenzamide, 5-Aziridino-2,4-dinitrobenzamide, 5-Aziridinyl-2,4-dinitrobenzamide, CB 1954, NSC 115829
IUPAC Name 5-(aziridin-1-yl)-2,4-dinitrobenzamide
SMILES NC(=O)c1cc(N2CC2)c(cc1[N+](=O)[O-])[N+](=O)[O-]

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C9H8N4O5/c10-9(14)5-3-7(11-1-2-11)8(13(17)18)4-6(5)12(15)16/h3-4H,1-2H2,(H2,10,14)
Formula Weight 252.0495
Chemical Name or Material Tretazicar
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.