Learn More
TREX1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-68672
 View more versions of this product
Product Code. 18681958
  • € 624.00 / 100µL
Description
TREX1 Polyclonal antibody specifically detects TREX1 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TREX1
Polyclonal
Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL
3'-5' exonuclease TREX1, AGS1, Aicardi-Goutieres syndrome 1, CRV, DKFZp434J0310, DNase III, DRN3, EC 3.1.11.2, HERNS, three prime repair exonuclease 1,3' repair exonuclease 1
This antibody was developed against a recombinant protein corresponding to amino acids: VDAHARPFGTIRPMYGVTASARTKPRPSAVTTTAHLATTRNTSPSLGESRGTKDLPPVKDPGALSRE
100 μg
DNA Repair, Editing and Processing Endonucleases
11277
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Immunofluorescence
Unconjugated
PBS (pH 7.2) and 40% Glycerol
Rabbit
Protein A purified