missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRF-2 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB110-57130CL1
This item is not returnable.
View return policy
Description
TRF-2 Polyclonal antibody specifically detects TRF-2 in Chinese Hamster,Human,Mouse,Primate,Rat samples. It is validated for Flow Cytometry,ELISA,Immunohistochemistry,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin),Dot Blot,Simple Western,ChIP assay (ChIP),KnockDown,Immunocytochemistry/Immunofluorescence
Specifications
| TRF-2 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Chinese Hamster, Human, Mouse, Primate, Rat | |
| Purified |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| CoraFluor 1 | |
| Telomeric DNA-binding protein, telomeric repeat binding factor 2, Telomeric repeat binding protein 2, telomeric repeat-binding factor 2, TERF2, TRBF2, TRF2, TTAGGG repeat binding factor 2, TTAGGG repeat-binding factor 2 | |
| This TRF-2 Antibody was developed against Baculovirus purified TRF2 protein. | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Transcription Factors and Regulators | |
| 7014 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction