Learn More
Invitrogen™ TRF1 Monoclonal Antibody (3H11)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA146089
This item is not returnable.
View return policy
Description
Suggested positive control: antigen standard for TERF1 (transient overexpression lysate).
This gene encodes a telomere specific protein which is a component of the telomere nucleoprotein complex. This protein is present at telomeres throughout the cell cycle and functions as an inhibitor of telomerase, acting in cis to limit the elongation of individual chromosome ends. The protein structure contains a C-terminal Myb motif, a dimerization domain near its N-terminus and an acidic N-terminus. Two transcripts of this gene are alternatively spliced products.
Specifications
| TRF1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| P54274 | |
| TERF1 | |
| Human TRF-1. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 3H11 | |
| Unconjugated | |
| TERF1 | |
| FLJ41416; hTRF1-AS; NIMA-interacting protein 2; PIN2; Telomeric protein Pin2/TRF1; telomeric repeat binding factor (NIMA-interacting) 1; telomeric repeat binding factor 1; telomeric repeat-binding factor 1; TERF1; TRBF1; TRF; TRF1; TTAGGG repeat-binding factor 1; t-TRF1 | |
| Mouse | |
| Protein A | |
| RUO | |
| 7013 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |