Learn More
Invitrogen™ TRF2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116069
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total TRF2.
TRF2 is a ubiquitously expressed protein that is implicated in the control of telomere length. TRF2, like TRF1 contains a Myb-related DNA binding motif. It binds to duplex TTAGGG repeats and is localized to all human telomeres in metaphase chromosomes. TRF2 is thought to protect chromosome ends by maintaining the correct structure at telomere termini. The use of mutant forms of TRF2 has implicated a role for TRF2 in the prevention of senescence in primary human cells. Recently, it has been shown that inhibition of TRF2 resulted in apoptosis in a subset of mammalian cell types.
Specifications
| TRF2 | |
| Polyclonal | |
| Unconjugated | |
| TERF2 | |
| Telomeric DNA-binding protein; telomeric repeat binding factor 2; telomeric repeat binding protein 2; telomeric repeat-binding factor 2; TERF2; TRBF2; TRF2; TTAGGG repeat-binding factor 2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 7014 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| Q15554 | |
| TERF2 | |
| A synthesized peptide derived from human TERF2(Accession Q15554), corresponding to amino acid residues G339-G389. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |