missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ tri-GalNAc-PEG3-Azide

Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Description
tri-GalNAc-PEG3-Azide is a functionalized asialoglycoprotein receptor (ASGPR) ligand for lysosomal targeting chimera (LYTAC) research and development; each molecule incorporates three ASGPR ligands with a PEG3 linker and azide group reactive handle ready for conjugation. Upon binding to the ASGPR, tri-GalNAc conjugates are efficiently internalized via ASGPR-mediated endocytosis. tri-GalNAc conjugation can be employed as a strategy to effectively deliver cargo such as RNA or Cas9 complexes in a cell-specific manner to hepatocytes. Can be used to generate LYTACs, or labeled with dye for tissue imaging.
Specifications
Specifications
| CAS | 2925590-71-0 |
| Quantity | 1 mg |
| Target | Degrader Building Blocks |
| Molecular Formula | C70H125N13O31 |
| Purity | 0.9 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction