Learn More
Tri-n-octylchlorotin, TRC
Specifications
Specifications
| Chemical Name or Material | Trioctyltin chloride |
| CAS | 2587-76-0 |
| Molecular Formula | C24 H51 Cl Sn |
| Quantity | 50 mg |
| Synonym | Trioctyltin chloride, Stannane','chlorotrioctyl-, Tin','chlorotrioctyl- (7CI), Trioctyltin chloride (6CI), Chlorotrioctylstannane, Chlorotrioctyltin, NSC 65479, Tri-n-octyltin chloride, Trioctylchlorostannane |
| InChI Formula | InChI=1S/3C8H17.ClH.Sn/c3*1-3-5-7-8-6-4-2;;/h3*1,3-8H2,2H3;1H;/q;;;;+1/p-1 |
| SMILES | CCCCCCCC[Sn](Cl)(CCCCCCCC)CCCCCCCC |
| IUPAC Name | chloro(trioctyl)stannane |
| Molecular Weight (g/mol) | 493.82 |
| Formula Weight | 494.2701 |