Learn More
Tri(Propylene Glycol Butyl Ether (Mixture of isomers), TRC
Product Code. 30413735
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30413735 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30413735 Supplier Toronto Research Chemicals Supplier No. TRCT1849355ML
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1-[1-(1-butoxypropan-2-yloxy)propan-2-yloxy]propan-2-ol
CAS 55934-93-5
Molecular Formula C13 H28 O4
Quantity 5 mL
Synonym Tri(Propylene Glycol Butyl Ether (Mixture of isomers), [2-(2-Butoxymethylethoxy)methylethoxy]propanol, 2-(2-Butoxymethylethoxy)methylethoxy)propanol, BFTG, Dalpad C, Dowanol TPnB, TPnB, Tripropylene Glycol Monobutyl Ether, Tripropylene Glycol n-Butyl Ether, Tripropyleneglycol Butyl Ether
InChI Formula InChI=1S/C13H28O4/c1-5-6-7-15-9-12(3)17-10-13(4)16-8-11(2)14/h11-14H,5-10H2,1-4H3
SMILES CCCCOCC(C)OCC(C)OCC(C)O
IUPAC Name 1-[1-(1-butoxypropan-2-yloxy)propan-2-yloxy]propan-2-ol
Molecular Weight (g/mol) 248.359
Formula Weight 248.199