missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Triadin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5119478
This item is not returnable.
View return policy
Description
Triadin Polyclonal Antibody for Western Blot, IHC (P)
The junction between the transverse tubules (T-tubules) and the sarcoplasmic reticulum (SR) of skeletal muscle is called the triad. At the triad, dihydropyridine receptors (DHPR's) of the T-tubule serve as voltage sensors in excitation-contraction coupling, while ryanodine receptors (RyR's), the calcium release channels, exist in the membrane of the terminal cisternae of the SR. It is thought that during slow phase depolarization of the T-tubule, a third protein, triadin (MW 95 kDa) transmits electrochemical signals to the SR through direct interaction with both DHPR's and RyR's. Though its exact role in this signaling process is unclear, triadin has been shown to co-localize with both DHPR and RYR at the junctional face of the terminal cisternae.
Specifications
| Triadin | |
| Polyclonal | |
| Unconjugated | |
| TRDN | |
| 2310045H21Rik; cardiac triadin; cardiac triadin isoform 1; cardiac triadin isoform 2; cardiac triadin isoform 3; CPVT5; EG432451; junctional sarcoplasmic reticulum glycoprotein; TDN; Trdn; Triadin; triadin 1; triadin 32 kDa (TRISK 32); triadin 49 kDa (TRISK 49); triadin 95; TRISK | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 59299 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q9QX75 | |
| TRDN | |
| Recombinant protein Triadin. The antigen corresponds to amino acid range 486-687 of the target protein. | |
| 200 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction