missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triamcinolone acetonide, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Triamcinolone acetonide |
| CAS | 76-25-5 |
| Analyte or Component Names | Triamcinolone acetonide |
| Shipping Condition | +20°C |
| Molecular Formula | C24 H31 F O6 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Triamcinolone Hexacetonide Imp. A (EP), Triamcinolone Acetonide, Pregna-1,4-diene-3,20-dione, 9-fluoro-11,21-dihydroxy-16,17-[(1-methylethylidene)bis(oxy)]-, (11beta,16alpha)-, Pregna-1,4-diene-3,20-dione, 9-fluoro-11beta,16alpha,17,21-tetrahydroxy-, cyclic 16,17-acetal with acetone (6CI,8CI), (11beta,16alpha)-9-Fluoro-11,21-dihydroxy-16,17-[(1-methylethylidene)bis(oxy)]pregna-1,4-diene-3,20-dione, 9-Fluoro-11beta,16alpha,17,21-tetrahydroxypregna-1,4-diene-3,20-dione-16,17-acetonide, 9-Fluoro-11beta,21-dihydroxy-16alpha,17-(isopropylidenedioxy)pregna-1,4-diene-3,20-dione, 9-Fluoro-11beta,21-dihydroxy-16alpha,17alpha-(isopropylidenedioxy)pregna-1,4-diene-3,20-dione, 9alpha-Fluoro-11beta,21-dihydroxy-16alpha,17alpha-isopropylidenedioxy-1,4-pregnadiene-3,20-dione, 9alpha-Fluoro-16alpha-hydroxyprednisolone 16alpha,17alpha-acetonide, Adcortyl, Adcortyl A, Aftach, Aristicort, Aristocort A, Aristocort acetonide, Aristoderm, Azmacort, Berlicort, Delphicort, Extracort, Ftorocort, I-Vation, Kenacort 40, Kenacort 80, Kenacort A, Kenacort A 40, Kenalog, Kenalog 10, Kenalog 40, Ledercort Cream, Lichtena, MaQuaid, NSC 21916, Nasacort, Omcilon A, Paralen, Pol'kortolon, Queyanshusong A, Respicort, Rheudenolone, Rineton, Solodelf, TAC 3, TAC 40, Tamceton, Tramacin, Tri-Nasal, Triacet, Triakort, Triam, Triamcinolone 16,17-acetonide, Triamcinolone 16alpha,17alpha-acetonide, Triamcort-Depot, Triamhexal, Triamonide 40, Trianopollon, Tricinolon, Triesence, Trivaris, Trymex, Valon A 10, Vetalog, Vetazine, Volon A, Volonimat, 9-Fluoro-11beta,21-dihydroxy-16alpha,17-(1-methylethylidenedioxy)pregna-1,4-diene-3,20-dione, Fluocinolone Acetonide Impurity H, Triamcinolone Hexacetonide Impurity A |
| InChI Formula | InChI=1S/C24H31FO6/c1-20(2)30-19-10-16-15-6-5-13-9-14(27)7-8-21(13,3)23(15,25)17(28)11-22(16,4)24(19,31-20)18(29)12-26/h7-9,15-17,19,26,28H,5-6,10-12H2,1-4H3/t15-,16-,17-,19+,21-,22-,23-,24+/m0/s1 |
| SMILES | CC1(C)O[C@@H]2C[C@H]3[C@@H]4CCC5=CC(=O)C=C[C@]5(C)[C@@]4(F)[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)CO |
| IUPAC Name | 9α-Fluoro-11β,21-dihydroxy-16α,17α-isopropylidenedioxypregna-1,4-diene-3,20-dione |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?