missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIB1 Antibody (OTI8C8), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74596H
This item is not returnable.
View return policy
Description
TRIB1 Monoclonal specifically detects TRIB1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| TRIB1 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| TRIB1 | |
| Full length recombinant protein of human TRIB1 (NP_079471) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI8C8 | |
| Western Blot, Immunohistochemistry | |
| C8FWG-protein-coupled receptor induced protein, GIG-2, GIG2G-protein-coupled receptor-induced protein 2, G-protein-coupled receptor-induced gene 2 protein, SKIP1, TRB-1, TRB1phosphoprotein regulated by mitogenic pathways, tribbles homolog 1, tribbles homolog 1 (Drosophila), tribbles-like protein 1 | |
| Mouse | |
| Affinity Purified | |
| Protein Kinase | |
| 10221.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction