missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIB2 Antibody (OTI8D11), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74597FR
This item is not returnable.
View return policy
Description
TRIB2 Monoclonal specifically detects TRIB2 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| TRIB2 | |
| Monoclonal | |
| DyLight 680 | |
| PBS with 0.05% Sodium Azide | |
| TRIB2 | |
| Full length recombinant protein of human TRIB2 (NP_067675) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| IgG2b |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI8D11 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| C5FW, FLJ57420, GS3955, TRB-2, TRB2tribbles homolog 2, tribbles homolog 2 (Drosophila) | |
| Mouse | |
| Affinity Purified | |
| Protein Kinase | |
| 28951.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction