missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIB3 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-82067CL1
This item is not returnable.
View return policy
Description
TRIB3 Polyclonal antibody specifically detects TRIB3 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| TRIB3 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| C20orf97, dJ1103G7.3, neuronal cell death inducible putative kinase, Neuronal cell death-inducible putative kinase, NIPK, p65-interacting inhibitor of NF-kappaB, p65-interacting inhibitor of NF-kappa-B, SINK, SKIP3, TRB-3, TRB3chromosome 20 open reading frame 97, tribbles homolog 3, tribbles homolog 3 (Drosophila) | |
| TRIB3 antibody was raised against a 17 amino acid peptide near the center of human TRIB3. The immunogen is located within amino acids 100 - 150 of TRIB3. Amino Acid Squence: EPYARLPPHKHVARPTE | |
| 0.1 mL | |
| Apoptosis, Cancer, Hypoxia | |
| 57761 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction