missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIB3 Monoclonal Antibody (OTI3G4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA807611
This item is not returnable.
View return policy
Description
A novel inhibitor of cell division, Tribbles, blocks mitosis at a critical point in Drosophila morphogenesis. Recently, mammalian homologues of Tribbles (Trbs) have been identified that control mitogen-activated protein kinase (MAPK) activity. TRIB3 also known as TRB-3, SINK, and SKIP3 is a member of Trbs family. Overexpression of this protein inhibits NF-kB-dependent transcription induced by tumor necrosis factor (TNF) stimulation or its downstream signaling proteins but does not inhibit NF-kB translocation to the nucleus and binding to DNA. TRIB3/SKIP3 is also overexpressed in multiple human tumors and is regulated by hypoxia.Specifications
| TRIB3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q96RU7 | |
| TRIB3 | |
| Human recombit protein fragment corresponding to amino acids 1-200 of human TRIB3 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3G4 | |
| Unconjugated | |
| TRIB3 | |
| C20orf97, NIPK, SINK, SKIP3, TRB3 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 57761 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction