missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIB3 Monoclonal Antibody (OTI3G4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA807611S
This item is not returnable.
View return policy
Description
TRIB3 Monoclonal antibody specifically detects TRIB3 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| TRIB3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q5VTR2 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 56254 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3G4 | |
| Unconjugated | |
| RNF20 | |
| C20orf97; DJ1103G7.3; Ifld2; induced in fatty liver dystrophy 2; kinase; Neuronal cell death Inducible Putative Kinase; neuronal cell death-inducible putative kinase; Nipk; p65-interacting inhibitor of NF-kappaB; p65-interacting inhibitor of NF-kappa-B; SINK; SKIP3; Trb3; TRB-3; TRIB3; Tribbles 3; Tribbles homolog 3; tribbles homolog 3 (Drosophila); tribbles pseudokinase 3 | |
| Human recombinant protein fragment corresponding to amino acids 493-754 of human RNF20 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction