missing translation for 'onlineSavingsMsg'
Learn More

Tricarballylic acid, 98%

Product Code. p-7026218
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11479607

Brand: Thermo Scientific Alfa Aesar B22103.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 99-14-9 | C6H8O6 | 176.12 g/mol

It is used as an alternative to DMDHEU (formaldehyde releasing cross- linking agent).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an alternative to DMDHEU (formaldehyde releasing cross- linking agent).

Solubility
Soluble in water (500 mg/ml at 18°C), methanol, and ether (slightly).

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-14-9
Molecular Formula C6H8O6
Molecular Weight (g/mol) 176.12
MDL Number MFCD00002723
InChI Key KQTIIICEAUMSDG-UHFFFAOYSA-N
Synonym tricarballylic acid, 1,2,3-propanetricarboxylic acid, carballylic acid, tricarballylate, beta-carboxyglutaric acid, propane 1,2,3-tricarboxylic acid, unii-ra5qh2j020, 3-carboxyglutaric acid, .beta.-carboxyglutaric acid, 1,2,3-propanetricarboxylicacid
PubChem CID 14925
ChEBI CHEBI:45969
IUPAC Name propane-1,2,3-tricarboxylic acid
SMILES OC(=O)CC(CC(O)=O)C(O)=O

Specifications

Melting Point 157°C to 162°C
Quantity 5 g
Beilstein 1783567
Merck Index 14,9622
Solubility Information Soluble in water (500mg/ml at 18°C),methanol,and ether (slightly).
Formula Weight 176.12
Percent Purity 98%
Chemical Name or Material Tricarballylic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.