Learn More
Trichloroacetic anhydride, 95%, Thermo Scientific Chemicals
Catalog Number p-4440188
Click to view available options
Quantity:
25 g
100 g
4124-31-6
C4Cl6O3
308.76
MEFKFJOEVLUFAY-UHFFFAOYSA-N
trichloroacetic anhydride, acetic acid, trichloro-, anhydride, acetic acid, 2,2,2-trichloro-, 1,1'-anhydride, acmc-1aliy, perchloroacetic anhydride, bis trichloroacetic anhydride, trichloroacetic acid anhydride, 2,2,2-trichloroacetic anhydride, trichloroacetyl 2,2,2-trichloroacetate
20079
(2,2,2-trichloroacetyl) 2,2,2-trichloroacetate
C(=O)(C(Cl)(Cl)Cl)OC(=O)C(Cl)(Cl)Cl
4124-31-6
C4Cl6O3
308.76
MEFKFJOEVLUFAY-UHFFFAOYSA-N
trichloroacetic anhydride, acetic acid, trichloro-, anhydride, acetic acid, 2,2,2-trichloro-, 1,1'-anhydride, acmc-1aliy, perchloroacetic anhydride, bis trichloroacetic anhydride, trichloroacetic acid anhydride, 2,2,2-trichloroacetic anhydride, trichloroacetyl 2,2,2-trichloroacetate
20079
(2,2,2-trichloroacetyl) 2,2,2-trichloroacetate
C(=O)(C(Cl)(Cl)Cl)OC(=O)C(Cl)(Cl)Cl
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Density 1.6900g/mL
Boiling Point 139°C to 141°C (60.0 mmHg)
Packaging Glass Bottle
Linear Formula [CCl3C(=O)]2O
Quantity 25 g
Specific Gravity 1.69
Solubility Information Solubility in water: reacts
Formula Weight 308.76
Percent Purity 95%
Chemical Name or Material Trichloroacetic anhydride
compliance-icons
Product Identifier
  • Trichloroacetic anhydride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.