missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trichomonas Vaginalis 1 Mouse anti-Protozoa, Clone: 1306/156, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-62964-0.025MG
This item is not returnable.
View return policy
Description
Trichomonas Vaginalis 1 Monoclonal specifically detects Trichomonas Vaginalis 1 in Protozoa samples. It is validated for ELISA.
Specifications
| Trichomonas Vaginalis 1 | |
| Monoclonal | |
| 1.0 mg/ml | |
| ELISA 1:100-1:2000 | |
| T vaginalis, TV | |
| Native | |
| 0.025 mg | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| ELISA | |
| 1306/156 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Protozoa | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction