missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Trichomonas vaginalis Monoclonal Antibody (J37A)
GREENER_CHOICE

Product Code. 15382747 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15382747 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15382747 Supplier Invitrogen™ Supplier No. MA518274

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Purity is >90%. Recommended pairs for ELISA: MA5-18274 (capture) and MA5-18273 (conjugated, detection).

TRUSTED_SUSTAINABILITY

Specifications

Antigen Trichomonas vaginalis
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone J37A
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias t vaginalis; T. vaginalis
Host Species Mouse
Immunogen Trichomonas vaginalis native protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Protozoa
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.