missing translation for 'onlineSavingsMsg'
Learn More

Triethyl Orthoacetate, TRC

Product Code. 30451714
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30451714 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30451714 Supplier Toronto Research Chemicals Supplier No. TRCT776850500ML
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Triethyl Orthoacetate
CAS 78-39-7
Molecular Formula C8H18O3
Quantity 500 mL
Synonym 1,1,1-Triethoxyethane, Orthoacetic Acid Triethyl Ester, Ethyl Orthoacetate, NSC 5596
InChI Formula InChI=1S/C8H18O3/c1-5-9-8(4,10-6-2)11-7-3/h5-7H2,1-4H3
SMILES CCOC(C)(OCC)OCC
IUPAC Name 1,1,1-triethoxyethane
Molecular Weight (g/mol) 162.23
Formula Weight 162.1256
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.