missing translation for 'onlineSavingsMsg'
Learn More

Triethylene Glycol Dimethanesulfonate, TRC

Product Code. 30453535
Change view
Click to view available options
Quantity:
2 g
Unit Size:
2g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30453535 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30453535 Supplier Toronto Research Chemicals Supplier No. TRCT7765002G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Triethylene Glycol Dimethanesulfonate
CAS 80322-82-3
Molecular Formula C8H18O8S2
Quantity 2 g
Synonym 2,2'-[1,2-Ethanediylbis(oxy)]bis-ethanol 1,1'-Dimethanesulfonate, Dimesyl Triethylene Glycol, Triethylene Glycol Dimesylate
InChI Formula InChI=1S/C8H18O8S2/c1-17(9,10)15-7-5-13-3-4-14-6-8-16-18(2,11)12/h3-8H2,1-2H3
SMILES CS(=O)(=O)OCCOCCOCCOS(=O)(=O)C
IUPAC Name 2-[2-(2-methylsulfonyloxyethoxy)ethoxy]ethyl methanesulfonate
Molecular Weight (g/mol) 306.35
Formula Weight 306.0443
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.