missing translation for 'onlineSavingsMsg'
Learn More

Triforin, TRC

Product Code. 30452936
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30452936 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30452936 Supplier Toronto Research Chemicals Supplier No. TRCT79365550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Triforine
CAS 26644-46-2
Molecular Formula C10 H14 Cl6 N4 O2
Quantity 50 mg
Synonym N,N'-[1,4-Piperazinediylbis(2,2,2-trichloroethylidene)]bis[formamide], 1,4-Bis(1-formamido-2,2,2-trichloroethyl)piperazine, Asepta Funginex, Biformylchlorazin, CA 70203, CELA 50, CELA-W 524, Funginex, N,N'-Bis(1-formamido-2,2,2-trichloroethyl)piperazine, NSC 263493, Saprol, Triforin, Triforine, W 524
InChI Formula InChI=1S/C10H14Cl6N4O2/c11-9(12,13)7(17-5-21)19-1-2-20(4-3-19)8(18-6-22)10(14,15)16/h5-8H,1-4H2,(H,17,21)(H,18,22)
SMILES ClC(Cl)(Cl)C(NC=O)N1CCN(CC1)C(NC=O)C(Cl)(Cl)Cl
IUPAC Name N-[2,2,2-trichloro-1-[4-(2,2,2-trichloro-1-formamidoethyl)piperazin-1-yl]ethyl]formamide
Molecular Weight (g/mol) 434.96
Formula Weight 431.9248
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.