missing translation for 'onlineSavingsMsg'
Learn More

Triheneicosanoin, TRC

Product Code. 30424485
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30424485 25 mg 25mg
30418767 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30424485 Supplier Toronto Research Chemicals Supplier No. TRCT26536025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Triheneicosanoin
CAS 26536-14-1
Molecular Formula C66 H128 O6
Quantity 25 mg
Synonym 1,2,3-Propanetriyl Ester Heneicosanoic Acid
InChI Formula InChI=1S/C66H128O6/c1-4-7-10-13-16-19-22-25-28-31-34-37-40-43-46-49-52-55-58-64(67)70-61-63(72-66(69)60-57-54-51-48-45-42-39-36-33-30-27-24-21-18-15-12-9-6-3)62-71-65(68)59-56-53-50-47-44-41-38-35-32-29-26-23-20-17-14-11-8-5-2/h63H,4-62H2,1-3H3
SMILES O=C(OCC(OC(=O)CCCCCCCCCCCCCCCCCCCC)COC(=O)CCCCCCCCCCCCCCCCCCCC)CCCCCCCCCCCCCCCCCCCC
IUPAC Name 2,3-di(henicosanoyloxy)propyl henicosanoate
Molecular Weight (g/mol) 1017.719
Formula Weight 1016.971
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.