missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM Antibody (TRIM-04), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-485AF647
This item is not returnable.
View return policy
Description
TRIM Monoclonal specifically detects TRIM in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| TRIM | |
| Monoclonal | |
| Alexa Fluor 647 | |
| pp29/30, T cell receptor associated transmembrane adaptor 1, T cell receptor interacting molecule, T-cell receptor interacting molecule, T-cell receptor-associated transmembrane adapter 1, T-cell receptor-interacting molecule, TCRIM, TRIMHSPC062 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| The antibody TRIM-04 reacts with T cell receptor-interacting molecule (TRIM), a 30 kDa adaptor protein expressed by T cells. | |
| Store at 4C in the dark. |
| Western Blot, Flow Cytometry | |
| TRIM-04 | |
| Western Blot, Flow Cytometry | |
| TRAT1 | |
| Recombinant fragment, corresponding to amino acids 29-186 of Human TRIM. | |
| 0.1 mL | |
| Adaptive Immunity, Immunology | |
| 50852 | |
| Human | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction