missing translation for 'onlineSavingsMsg'
Learn More

TRIM2 Antibody (OTI2B10), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-74606AF594

Product Code. 18134677

  • € 751.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

TRIM2 Monoclonal specifically detects TRIM2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TRIM2
Monoclonal
Alexa Fluor 594
50 mM sodium borate with 0.05% sodium azide
TRIM2
Recombinant protein fragment corresponding to amino acids 1-100 and 1500-1600 of human TRIM2 (NP_775105) produced in HEK293T cells.
Primary
Human, Mouse, Rat
IgG1
Western Blot, Flow Cytometry
OTI2B10
Western Blot, Flow Cytometry
KIAA0517tripartite motif protein 2, RING finger protein 86, RNF86tripartite motif protein TRIM2, tripartite motif containing 2, tripartite motif-containing 2, tripartite motif-containing protein 2
Mouse
0.1 mL
23321
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.