missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM2 Antibody (OTI2B10), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-74606CL1
This item is not returnable.
View return policy
Description
TRIM2 Monoclonal antibody specifically detects TRIM2 in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Western Blot,CyTOF
Specifications
| TRIM2 | |
| Monoclonal | |
| CoraFluor 1 | |
| KIAA0517tripartite motif protein 2, RING finger protein 86, RNF86tripartite motif protein TRIM2, tripartite motif containing 2, tripartite motif-containing 2, tripartite motif-containing protein 2 | |
| Human recombinant protein fragment corresponding to amino acids 1-100 and 1500-1600 of human SHPRH (NP_775105) produced in HEK293T cell. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Flow Cytometry, Western Blot, CyTOF | |
| OTI2B10 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 23321 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction