missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRIM2 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA518152
This item is not returnable.
View return policy
Description
This antibody is predicted to react with bovine and canine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.
The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to cytoplasmic filaments. Its function has not been identified.
Specifications
| TRIM2 | |
| Polyclonal | |
| Unconjugated | |
| Trim2 | |
| CMT2R; E3 ubiquitin-protein ligase TRIM2; KIAA0517; mKIAA0517; narf; Neural activity-related RING finger protein; RING finger protein 86; RING-type E3 ubiquitin transferase TRIM2; RNF86; TRIM2; tripartite motif containing 2; tripartite motif protein 2; tripartite motif protein TRIM2; tripartite motif-containing 2; tripartite motif-containing protein 2 | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 23321, 361970 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| D3ZQG6, Q9C040 | |
| Trim2 | |
| Synthetic peptide sequence (DSGNHCFKVYRYLQ) corresponding to the C-terminus amino acids of TRIM2 (aa 731-744). | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction