missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRIM2 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142724
This item is not returnable.
View return policy
Description
Additional Information: This antibody is expected to recognize both reported isoforms (NP_056086.2; NP_001123539.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:16,000.
The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to cytoplasmic filaments. Its function has not been identified.
Specifications
| TRIM2 | |
| Polyclonal | |
| Unconjugated | |
| Trim2 | |
| CMT2R; E3 ubiquitin-protein ligase TRIM2; KIAA0517; mKIAA0517; narf; Neural activity-related RING finger protein; RING finger protein 86; RING-type E3 ubiquitin transferase TRIM2; RNF86; TRIM2; tripartite motif containing 2; tripartite motif protein 2; tripartite motif protein TRIM2; tripartite motif-containing 2; tripartite motif-containing protein 2 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 80890 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q9ESN6 | |
| Trim2 | |
| Peptide with sequence C-STFDELQKTLNVRKS. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction