missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trim22 Antibody (OTI1D11), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74608AF647
This item is not returnable.
View return policy
Description
Trim22 Monoclonal specifically detects Trim22 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Trim22 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| TRIM22 | |
| Recombinant protein fragment corresponding to amino acids 61-406 of human Trim22 (NP_006065) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI1D11 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| 50 kDa-stimulated trans-acting factor, E3 ubiquitin-protein ligase TRIM22, EC 6.3.2, EC 6.3.2.-, GPSTAF50, RING finger protein 94, RNF94stimulated trans-acting factor (50 kDa), Staf-50, STAF50staf-50, tripartite binding motif 22, tripartite motif containing 22, tripartite motif protein TRIM22, tripartite motif-containing 22, Tripartite motif-containing protein 22 | |
| Mouse | |
| Affinity Purified | |
| Zinc Finger | |
| 10346.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction