missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM24 Monoclonal Antibody (OTI3A8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA802789
This item is not returnable.
View return policy
Description
The protein encoded by this gene mediates transcriptional control by interaction with the activation function 2 (AF2) region of several nuclear receptors, including the estrogen, retinoic acid, and vitamin D3 receptors. The protein localizes to nuclear bodies and is thought to associate with chromatin and heterochromatin-associated factors. The protein is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains - a RING, a B-box type 1 and a B-box type 2 - and a coiled-coil region. Two alternatively spliced transcript variants encoding different isoforms have been described for this gene.Specifications
| TRIM24 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O15164 | |
| TRIM24 | |
| Human recombit protein fragment corresponding to amino acids 706-1016 of human TRIM24 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI3A8 | |
| Unconjugated | |
| TRIM24 | |
| PTC6, RNF82, TF1A, TIF1, TIF1A, TIF1ALPHA, hTIF1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 8805 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction