missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM25 Antibody (5B5B10) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 203.00 - € 552.00
Specifications
| Antigen | TRIM25 |
|---|---|
| Clone | 5B5B10 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:50-1:250, Immunohistochemistry-Paraffin 1:200-1:1000 |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18239082
|
Novus Biologicals
NBP2-61820 |
0.1 mL |
€ 552.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18633508
|
Novus Biologicals
NBP2-61820-0.025ml |
0.025 mL |
€ 203.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TRIM25 Monoclonal specifically detects TRIM25 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| TRIM25 | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| Cancer, Zinc Finger | |
| 7706 | |
| Purified recombinant fragment of human TRIM25 (AA: 211-360) expressed in E. Coli. | |
| Primary | |
| 26.4 kDa |
| 5B5B10 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:50-1:250, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| Unconjugated | |
| Mouse | |
| EC 6.3.2.19, EC 6.3.2.n3, EFPZ147, Estrogen-responsive finger protein, RING finger protein 147, RNF147E3 ubiquitin/ISG15 ligase TRIM25, tripartite motif containing 25, tripartite motif-containing 25, Tripartite motif-containing protein 25, Ubiquitin/ISG15-conjugating enzyme TRIM25, Zinc finger protein 147, zinc finger protein 147 (estrogen-responsive finger protein), zinc finger protein-147, ZNF147tripartite motif protein TRIM25 | |
| TRIM25 | |
| IgG2b | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title