missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM25 Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76826DL594
This item is not returnable.
View return policy
Description
TRIM25 Polyclonal antibody specifically detects TRIM25 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| TRIM25 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| DyLight 594 | |
| EC 6.3.2.19, EC 6.3.2.n3, EFPZ147, Estrogen-responsive finger protein, RING finger protein 147, RNF147E3 ubiquitin/ISG15 ligase TRIM25, tripartite motif containing 25, tripartite motif-containing 25, Tripartite motif-containing protein 25, Ubiquitin/ISG15-conjugating enzyme TRIM25, Zinc finger protein 147, zinc finger protein 147 (estrogen-responsive finger protein), zinc finger protein-147, ZNF147tripartite motif protein TRIM25 | |
| Antibody was raised against a 15 amino acid synthetic peptide from near the amino terminus of human TRIM25. The immunogen is located within amino acids 40 - 90 of TRIM25. Amino Acid Squence: CRAVYQARPQLHKNT | |
| 0.1 mL | |
| Cancer, Zinc Finger | |
| 7706 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction