missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM26 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-47595
This item is not returnable.
View return policy
Description
TRIM26 Polyclonal specifically detects TRIM26 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| TRIM26 | |
| Polyclonal | |
| Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Acid finger protein, AFP, RNF95RING finger protein 95, tripartite motif containing 26, tripartite motif-containing protein 26, widely expressed acid zinc finger protein, Zinc finger protein 173, ZNF173tripartite motif-containing 26 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| TRIM26 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: FKKENIRPVWQLASLVENIERLKVDKGRQPGEVTREQQDAKLCERHREKLHYYCEDDGKLLCVM | |
| 0.1 mL | |
| Zinc Finger | |
| 7726 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction