missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRIM26 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA540836
This item is not returnable.
View return policy
Description
Peptide sequence: KKLQDQRQYI VAEFEQGHQF LREREEHLLE QLAKLEQELT EGREKFKSRG Sequence homology: Cow: 100%; Dog: 100%; Horse: 85%; Human: 100%; Mouse: 92%; Pig: 85%; Rabbit: 100%; Rat: 92%.
TRIM26 is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to cytoplasmic bodies. Although the function of the protein is unknown, the RING domain suggests that the protein may have DNA-binding activity. The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to cytoplasmic bodies. Although the function of the protein is unknown, the RING domain suggests that the protein may have DNA-binding activity. The gene localizes to the major histocompatibility complex (MHC) class I region on chromosome 6.
Specifications
| TRIM26 | |
| Polyclonal | |
| Unconjugated | |
| TRIM26 | |
| Acid finger protein; AFP; AI462198; RING finger protein 95; RNF95; TRIM26; tripartite motif containing 26; tripartite motif protein 26; tripartite motif protein TRIM26; tripartite motif-containing 26; tripartite motif-containing protein 26; widely expressed acid zinc finger protein; Zfp173; Zfp1736; zinc finger protein 173; Znf173 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 7726 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q12899 | |
| TRIM26 | |
| Synthetic peptide directed towards the middle region of human TRIM26. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction