missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM27 Antibody (OTI4D4), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-46214
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry
TRIM27 Monoclonal specifically detects TRIM27 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| TRIM27 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:2000, Immunohistochemistry 1:150, Immunohistochemistry-Paraffin 1:150 | |
| Ret finger proteinRING finger protein 76, RFPtripartite motif protein TRIM27, RNF76RFP transforming protein, tripartite motif containing 27, tripartite motif-containing 27, Tripartite motif-containing protein 27, zinc finger protein RFP | |
| Mouse | |
| 58.3 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| OTI4D4 | |
| Unconjugated | |
| PBS (pH 7.3), 1.0% BSA, 50% Glycerol with 0.02% Sodium Azide | |
| TRIM27 | |
| Full length human recombinant protein of human TRIM27 (NP_006501) produced in E.coli. | |
| Immunogen affinity purified | |
| RUO | |
| 5987 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction